Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluorotoluene, TCI America™
SDP

Catalog No. D235110G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D235110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D235110G Supplier TCI America Supplier No. D235110G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 452-76-6
Molecular Formula C7H6F2
Molecular Weight (g/mol) 128.122
MDL Number MFCD00000332
InChI Key MPXDAIBTYWGBSL-UHFFFAOYSA-N
Synonym 2,4-difluorotoluene, difluorotoluene, benzene, 2,4-difluoro-1-methyl, 2,4-difluorotolune, 2, 4-difluorotoluene, 2,4-difluoro methyl benzene, 2,4-difluoro-1-methyl-benzene, 2,4-difluor-1-methylbenzol, pubchem1612, pubchem1613
PubChem CID 67983
IUPAC Name 2,4-difluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)F

Specifications

Color Yellow
Boiling Point 116°C
Quantity 10 g
UN Number 1993
Formula Weight 128.12
Physical Form Liquid
Chemical Name or Material 2,4-Difluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.