Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-dihydro-5-(4-nitrophenyl)-2-phenyl-3H-pyrazol-3-one | 34320-83-7 | MFCD00506084 | 99.7% | 281.27 | C15H11N3O3 | 10 MG
SDP

Catalog No. 5000240307
Encompass_Preferred

TCS PrP Inhibitor 13 is a small-molecule antiprion agent that inhibits cellular prion protein (PrPC) and reduces protease-resistant prion accumulation. It is supplied as a solid for research use and shows potent activity in cellular assays (IC50 3 nM).

  • Chemical formula: C15H11N3O3.
  • Molecular weight: 281.27.
  • Purity: 99.7%.
  • Appearance: solid, white to yellow.
  • Storage conditions: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Reported activity: IC50 3 nM in ScN2a and F3 cells.
  • Biological effect: induces apoptosis in schwannoma cells.

Catalog No. 50-002-40307 Supplier Medchemexpress LLC Supplier No. HY10766210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCS PrP Inhibitor 13 is a small-molecule antiprion agent that inhibits cellular prion protein (PrPC) and reduces protease-resistant prion accumulation. It is supplied as a solid for research use and shows potent activity in cellular assays (IC50 3 nM).

  • Chemical formula: C15H11N3O3.
  • Molecular weight: 281.27.
  • Purity: 99.7%.
  • Appearance: solid, white to yellow.
  • Storage conditions: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Reported activity: IC50 3 nM in ScN2a and F3 cells.
  • Biological effect: induces apoptosis in schwannoma cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.