Learn More
Selleck Chemical LLC 2.4-Dihydroxyacetophenone S4762-25mg
SDP
Catalog No. 503109679
Encompass_Preferred
2 4-Dihydroxyacetophenone (Resacetophenone 4-Acetylresorcinol) is a plant metabolite that can act as a qualitative reagent for ferric iron
Catalog No. 50-310-9679 Supplier Selleck Chemical LLC Supplier No. S476225MG
2 4-Dihydroxyacetophenone (Resacetophenone 4-Acetylresorcinol) is a plant metabolite that can act as a qualitative reagent for ferric iron