Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Dihydroxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. D0561500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0561500G 500 g
D056125G 25 g
D0561100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0561500G Supplier TCI America Supplier No. D0561500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89-84-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002279
InChI Key SULYEHHGGXARJS-UHFFFAOYSA-N
Synonym 2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
PubChem CID 6990
ChEBI CHEBI:18414
IUPAC Name 1-(2,4-dihydroxyphenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(O)C=C1O

Specifications

Melting Point 145°C
Quantity 500 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,4′-Dihydroxyacetophenone
EINECSNumber (3)-1250
RTECSNumber AM7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.