Learn More
Medchemexpress LLC 3-Color Prestained Protein Marker (10-190 kDa) 
SDP
Catalog No. 501970211
Encompass_Preferred
MCE 3-Color Prestained Protein Marker has 8 blue-stained bands and 2 reference bands (one green and one orange band at 23 kDa and 65 kDa respectively). This Protein Marker is designed for monitoring protein separation during SDS-PAGE, confirming Western blot transfer efficiency on membranes (PVDF or nitrocellulose membrane) and for approximating the size of target proteins on the gel or blot.MCE 3-Color Prestained Protein Marker is supplied in gel loading buffer and is ready to use, with no heating, diluting or additional reducing agent necessary.
Catalog No. 50-197-0211 Supplier Medchemexpress LLC Supplier No. HYK10111 ML
MCE 3-Color Prestained Protein Marker has 8 blue-stained bands and 2 reference bands (one green and one orange band at 23 kDa and 65 kDa respectively). This Protein Marker is designed for monitoring protein separation during SDS-PAGE, confirming Western blot transfer efficiency on membranes (PVDF or nitrocellulose membrane) and for approximating the size of target proteins on the gel or blot.MCE 3-Color Prestained Protein Marker is supplied in gel loading buffer and is ready to use, with no heating, diluting or additional reducing agent necessary.