Learn More
Selleck Chemical LLC 2 4-Dihydroxybenzoic acid
SDP
Catalog No. 5000003634
Encompass_Preferred
2 4-Dihydroxybenzoic acid ( -resorcylic acid p-Hydroxysalicylic acid) is a degradation product of cyanidin glycosides from tart cherries in cell cultures and a metabolite found in human plasma after cranberry juice consumption
Catalog No. 50-000-03634 Supplier Selleck Chemical LLC Supplier No. S629325MG
2 4-Dihydroxybenzoic acid ( -resorcylic acid p-Hydroxysalicylic acid) is a degradation product of cyanidin glycosides from tart cherries in cell cultures and a metabolite found in human plasma after cranberry juice consumption