Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2,4-Dihydroxybutyric acid lithium salt, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111013207 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3207 50 mg
11-101-3206 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3207 Supplier MilliporeSigma Supelco Supplier No. 8992150MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym 2,4-Dihydroxybutanoic acid lithium salt

Specifications

CAS 1518-62-3
Linear Formula C4H8O4
Quantity 50 mg
Formula Weight 120.10
Percent Purity ≥95.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.