Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dihydroxydiphenylmethane 98.0+%, TCI America™
SDP

Catalog No. D19401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D19401G 1 g
D19405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D19401G Supplier TCI America Supplier No. D19401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2467-03-0
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.24
MDL Number MFCD00060113
InChI Key LVLNPXCISNPHLE-UHFFFAOYSA-N
PubChem CID 75576
IUPAC Name 4-[(2-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(CC2=CC=CC=C2O)C=C1

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Dihydroxydiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.