Learn More
Sigma Aldrich 2,4-Dimethoxy-5-fluoronitrobenzene
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 6-heptynoic acid is an alkynoic acid with an acetylene bond. it undergoes condensation with various pyrroles to afford optical diverse fluorescent dyes with a terminal alkyne.6-heptynoic acid may be used for the following syntheses:
- Alkyne functionalized boradiazaindacenes (bodipy)dyes
- Natural products epothilone b and d
- Hymenialdisine (hmd) and aldisine (ad) affinity resins
- Alkynyl esters
Specifications
Specifications
| CAS | 195136-62-0 |
| Molecular Formula | C8H8FNO4 |
| Linear Formula | C8H8FNO4 |
| MDL Number | MFCD03789118 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 201.15 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.