Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxybenzaldoxime 95.0+%, TCI America™
SDP

Catalog No. D48515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48515G Supplier TCI America Supplier No. D48515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 31874-34-7
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD00082783
InChI Key SFDRVCQSVTYHLU-UXBLZVDNSA-N
Synonym 2,4-dimethoxybenzaldoxime, 2,4-dimethoxybenzaldehyde oxime, e-n-2,4-dimethoxyphenyl methylidene hydroxylamine, benzaldehyde, 2,4-dimethoxy-, oxime, 2.4-dimethoxybenzaldoxim, e-2,4-dimethoxybenzaldehyde oxime, 2,4-dimethoxyphenyl hydroxyimino methane, ne-n-2,4-dimethoxyphenyl methylidene hydroxylamine
PubChem CID 6871293
IUPAC Name (NE)-N-[(2,4-dimethoxyphenyl)methylidene]hydroxylamine
SMILES COC1=CC(=C(C=C1)C=NO)OC

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxybenzaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.