Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dimethoxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. D28905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28905G Supplier TCI America Supplier No. D28905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5449-69-4
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD01311508
InChI Key QWWJLMQOKZOTNX-UHFFFAOYSA-N
PubChem CID 226205
IUPAC Name (2-methoxyphenyl)-(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C2=CC=CC=C2OC

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Dimethoxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.