Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dimethoxybenzylamine

Catalog No. 4327255ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-272-55ML 5 mL
43-272-525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 43-272-55ML Supplier Sigma Aldrich Supplier No. 4327255ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4-dimethoxybenzylamine can be preprepared by reduction (nabh4 , bf3.oet2, thf) of 2,4-dimethoxybenzonitrile.2,4-dimethoxybenzylamine is an amine nucleophile used to investigate the 1,4- reactivity of 5-bromo-2-indene-1-one. it may be used in the following studies:
  • As an ammonia equivalent in the concise synthesis of a series of 2,4,5-trisubstituted oxazoles, via a tandem ugi/robinson-gabriel reaction sequence
  • Total synthesis of (-)-muraymycin (mry) d2 and its epimer, the antibacterial nucleoside natural product
  • Two-step synthesis of amide derivatives of uracil polyoxin c (upoc) methyl ester using the ugi reaction
  • Synthesis of n-hydroxythiourea
  • Synthesis of anti-hiv-1 agents

Specifications

CAS 20781-20-8
Density 1.113 g/mL (at 25°C (literature))
Boiling Point 140°C (1 mmHg)
Molecular Formula C9H13NO2
Linear Formula (CH3O)2C6H3CH2NH2
Refractive Index n20/D 1.549 (literature)
MDL Number MFCD00052393
Quantity 5 mL
Molecular Weight (g/mol) 167.21
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.