Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxycinnamic Acid 96.0+%, TCI America™
SDP

Catalog No. D236425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D236425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D236425G Supplier TCI America Supplier No. D236425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6972-61-8
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00016837
InChI Key YIKHDPHTFYWYJV-GQCTYLIASA-N
Synonym 2,4-dimethoxycinnamic acid, 3-2,4-dimethoxyphenyl acrylic acid, 2',4'-dimethoxycinnamic acid, e-3-2,4-dimethoxyphenyl prop-2-enoic acid, 2e-3-2,4-dimethoxyphenyl acrylic acid, 3-2,4-dimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,4-dimethoxyphenyl, 2e-3-2,4-dimethoxyphenyl prop-2-enoic acid, dimethoxycinnamicacid, 3-2,4-dimethoxyphenyl-2-propenoic acid
PubChem CID 734009
IUPAC Name (2E)-3-(2,4-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(\C=C\C(O)=O)C(OC)=C1

Specifications

Melting Point 184°C
Quantity 25 g
Formula Weight 208.21
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethoxycinnamic Acid
RTECSNumber UD3334500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.