Learn More
Medchemexpress LLC 2,4-dimethoxypyrimidine-5-boronic acid | 89641-18-9 | MFCD01318985 | 99.7% | 183.96 g/mol | C6H9BN2O4 | 5 G
SDP
Catalog No. 5000302238
Encompass_Preferred
2,4-Dimethoxypyrimidine-5-boronic acid is a boronic-acid-containing reagent used as an organic building block and intermediate in medicinal chemistry and synthetic research. It is commonly employed in cross-coupling reactions and small-molecule synthesis to introduce boron-containing functionality for further derivatization.
  • High purity: 99.73%.
  • Defined molecular weight: 183.96 g/mol.
  • Chemical formula: C6H9BN2O4.
  • Suitable for Suzuki-Miyaura and other cross-coupling reactions.
  • Available in multiple pack sizes for research-scale use.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-003-02238 Supplier Medchemexpress LLC Supplier No. HYW0163125G
2,4-Dimethoxypyrimidine-5-boronic acid is a boronic-acid-containing reagent used as an organic building block and intermediate in medicinal chemistry and synthetic research. It is commonly employed in cross-coupling reactions and small-molecule synthesis to introduce boron-containing functionality for further derivatization.
  • High purity: 99.73%.
  • Defined molecular weight: 183.96 g/mol.
  • Chemical formula: C6H9BN2O4.
  • Suitable for Suzuki-Miyaura and other cross-coupling reactions.
  • Available in multiple pack sizes for research-scale use.
  • Certificate of analysis and safety data sheet available.