Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dimethyl-1-nitrobenzene

Catalog No. 171018100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-101-8100G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-101-8100G Supplier Sigma Aldrich Supplier No. 171018100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,4-Dimethyl-1-nitrobenzene

Specifications

CAS 89-87-2
Melting Point 7°C to 9°C (lit.)
Density 1.117 g/mL (at 25°C (literature))
Boiling Point 244°C (lit.)
Molecular Formula C8H9NO2
Refractive Index n20/D 1.549 (literature)
Linear Formula (CH3)2C6H3NO2
MDL Number MFCD00007169
Quantity 250 mg
Synonym 1,3-Dimethyl-4-nitrobenzene; 4-Nitro-m-xylene
Molecular Weight (g/mol) 151.16
Percent Purity 98%; 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.