Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-Dimethyl-1H-imidazole-5-carbaldehyde | 68282-52-0 | 99.9% | C6H8N2O | 100 MG
SDP

Catalog No. 5000434265
Encompass_Preferred

2,4-Dimethyl-1H-imidazole-5-carbaldehyde is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.

  • A drug intermediate for synthesis
  • Purity of 99.9%
  • Appears as a solid
  • Color ranges from light yellow to yellow
  • Molecular weight of 124.14
  • Chemical formula C6H8N2O

Catalog No. 50-004-34265 Supplier Medchemexpress LLC Supplier No. HY65067100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-Dimethyl-1H-imidazole-5-carbaldehyde is a drug intermediate used for the synthesis of various active compounds. It is intended for research use only and is not sold to patients.

  • A drug intermediate for synthesis
  • Purity of 99.9%
  • Appears as a solid
  • Color ranges from light yellow to yellow
  • Molecular weight of 124.14
  • Chemical formula C6H8N2O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.