Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-Dimethyl-1H-imidazole-5-carbaldehyde | 68282-52-0 | 99.9% | C6H8N2O | 250 MG
SDP

Catalog No. 5000358592
Encompass_Preferred

2,4-Dimethyl-1H-imidazole-5-carbaldehyde is a drug intermediate for the synthesis of various active compounds.

  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 124.14
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture)
  • Shipping at room temperature (continental US)

Catalog No. 50-003-58592 Supplier Medchemexpress LLC Supplier No. 250MGHY65067
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-Dimethyl-1H-imidazole-5-carbaldehyde is a drug intermediate for the synthesis of various active compounds.

  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 124.14
  • Store at -20°C, sealed, away from moisture
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (sealed storage, away from moisture)
  • Shipping at room temperature (continental US)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.