Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-2-pentanol, 96%, Thermo Scientific™

Catalog No. AAL0879503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0879503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0879503 Supplier Thermo Scientific Chemicals Supplier No. L0879503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-06-9
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00021811
InChI Key FMLSQAUAAGVTJO-UHFFFAOYSA-N
Synonym 2,4-dimethyl-2-pentanol, isobutyldimethylcarbinol, 2-pentanol, 2,4-dimethyl, acmc-20aoch, 2-pentanol,4-dimethyl, 2-pentanol,2,4-dimethyl, 2-pentanol, 2,4-dimethyl-8ci 9ci
PubChem CID 12235
IUPAC Name 2,4-dimethylpentan-2-ol
SMILES CC(C)CC(C)(C)O

Specifications

Density 0.81
Boiling Point 132°C to 134°C
Flash Point 38°C (100°F)
Refractive Index 1.417
Quantity 1 g
UN Number UN1987
Beilstein 1731623
Formula Weight 116.2
Percent Purity 96%
Chemical Name or Material 2,4-Dimethyl-2-pentanol
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 210-877-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.