Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-dimethyl-3-furaldehyde, Thermo Scientific™

Catalog No. p-4391323
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC65804CB 250 mg Amber Glass Bottle
CC65804DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC65804CB Supplier Thermo Scientific Chemicals Supplier No. CC65804CB
Only null left
Add to Cart

Chemical Identifiers

CAS 75002-34-5
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key MGSAYHDYMMHZQE-UHFFFAOYSA-N
Synonym 2,4-dimethyl-3-furaldehyde, 3-furancarbaldehyde,2,4-dimethyl, 3-furancarboxaldehyde,2,4-dimethyl, 3-furancarboxaldehyde, 2,4-dimethyl-9ci
PubChem CID 21702758
IUPAC Name 2,4-dimethylfuran-3-carbaldehyde
SMILES CC1=COC(=C1C=O)C

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 124.14
Percent Purity 95%
Chemical Name or Material 2, 4-dimethyl-3-furaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.