Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-5-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. D51961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51961G 1 g
D51965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51961G Supplier TCI America Supplier No. D51961G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2124-47-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00025199
InChI Key DUIOVGPUAJSYCD-UHFFFAOYSA-N
Synonym 5-Nitro-2,4-xylidine
PubChem CID 228007
IUPAC Name 2,4-dimethyl-5-nitroaniline
SMILES CC1=CC(=C(C=C1N)[N+](=O)[O-])C

Specifications

Melting Point 124°C
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dimethyl-5-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.