Learn More
Medchemexpress LLC 2,4-dimethylaniline-d6 | 1071170-27-8 | MFCD22565800 | 127.22 | C8H5D6N | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW65384910MG
Encompass_Preferred
2,4-Xylidine-d6 is a deuterium-labeled isotopologue of 2,4-xylidine (2,4-dimethylaniline) supplied in small quantities for laboratory research. It is used primarily as an internal standard or tracer for analytical methods, including mass spectrometry and chromatographic assays.
  • Deuterium-labeled internal standard for analytical methods.
  • High isotopic labeling for clear mass shifts in MS analyses.
  • Available in small vial quantities suitable for method development.
  • Molecular formula C8H5D6N; molecular weight 127.22.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84064