Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-dimethylaniline-d6 | 1071170-27-8 | MFCD22565800 | 99.0% | 127.22 | C8H5D6N | 5 MG
SDP

Catalog No. 5000284727
Encompass_Preferred

2,4-Xylidine-d6 is the deuterium-labeled analogue of 2,4-xylidine (2,4-dimethylaniline) used as a stable isotope standard and tracer in analytical chemistry and mass spectrometry. It is supplied for laboratory research use only.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • Molecular formula C8H5D6N; molecular weight 127.22.
  • CAS number 1071170-27-8.
  • Supplied in small preweighed quantities suitable for analytical standards.
  • High purity (≈99.0%) based on certificate of analysis.

Catalog No. 50-002-84727 Supplier Medchemexpress LLC Supplier No. HYW6538495MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,4-Xylidine-d6 is the deuterium-labeled analogue of 2,4-xylidine (2,4-dimethylaniline) used as a stable isotope standard and tracer in analytical chemistry and mass spectrometry. It is supplied for laboratory research use only.

  • Deuterium-labeled internal standard for mass spectrometry and tracer studies.
  • Molecular formula C8H5D6N; molecular weight 127.22.
  • CAS number 1071170-27-8.
  • Supplied in small preweighed quantities suitable for analytical standards.
  • High purity (≈99.0%) based on certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.