Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylhexane, Thermo Scientific™

Catalog No. AC469280050
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469280050 5 mL
AC469280010 1 mL
AC469280250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC469280050 Supplier Thermo Scientific Chemicals Supplier No. 469280050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 589-43-5
Molecular Formula C8H18
Molecular Weight (g/mol) 114.23
MDL Number 00039929
InChI Key HDGQICNBXPAKLR-UHFFFAOYNA-N
IUPAC Name 2,4-dimethylhexane
SMILES CCC(C)CC(C)C

Specifications

Alpha Vector DIMETHYLHEXANE
Melting Point 108°C to 109°C
Color Colorless
Flash Point 10°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Refractive Index 1.3940 to 1.3960 (20°C, 589 nm)
Quantity 5 mL
Solubility Information In water: (20°C) almost insoluble
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylhexane
Show More Show Less
Hazard Category Flame
Hazard Statement Exclamation mark
EINECSNumber 209-649-6
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.