Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyloxazole 98.0+%, TCI America™
SDP

Catalog No. D37661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37661G 1 g
D37665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37661G Supplier TCI America Supplier No. D37661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7208-05-1
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.12
MDL Number MFCD00234124
InChI Key PSOZJOZKEVZLKZ-UHFFFAOYSA-N
PubChem CID 138961
IUPAC Name 2,4-dimethyl-1,3-oxazole
SMILES CC1=COC(=N1)C

Specifications

Color Yellow
Boiling Point 108°C
Quantity 1 g
UN Number 1993
Formula Weight 97.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyloxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.