Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dimethylpyrrole

Catalog No. 3908361G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-083-61G 1 g
39-083-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-083-61G Supplier Sigma Aldrich Supplier No. 3908361G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4-dimethylpyrrole is a substituted pyrrole. its synthesis by using ethyl acetoacetate as starting material has been reported. its basicity has been evaluated from UV spectral data. its photodecomposition on irradiation has been reported to afford h2, ch4, c2h6 and polymeric products.2,4-dimethylpyrrole may be used in the synthesis of the following:
  • 2,5-bis(2α,4α-dimethyl-5α-pyrryl)p-benzoquinone
  • Boron dipyrromethene (bodipy) dyes
  • 2,4-dimethyl-6-methoxyprodigiosene

Specifications

CAS 625-82-1
Density 0.924 g/mL (at 25°C (literature))
Boiling Point 165°C to 167°C (lit.)
Molecular Formula C6H9N
Refractive Index n20/D 1.496 (literature)
Linear Formula C6H9N
MDL Number MFCD00192088
Quantity 1 g
Molecular Weight (g/mol) 95.14
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.