Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,4-Dimethylthiazole-5-carboxylic acid | 53137-27-2 | 157.19 | 100 MG
SDP

Catalog No. 5000449562
Encompass_Preferred

2,4-Dimethylthiazole-5-carboxylic acid is a building block utilized in chemical synthesis. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Molecular formula: C6H7NO2S
  • CAS number: 53137-27-2
  • Building block for chemical synthesis
  • For research use only

Catalog No. 50-004-49562 Supplier Medchemexpress LLC Supplier No. HYW002255100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2,4-Dimethylthiazole-5-carboxylic acid is a building block utilized in chemical synthesis. It is intended for research use only.

  • Solid appearance
  • White to off-white color
  • Molecular formula: C6H7NO2S
  • CAS number: 53137-27-2
  • Building block for chemical synthesis
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.