Learn More
Medchemexpress LLC 2,4-Dimethylthiazole-5-carboxylic acid | 53137-27-2 | 157.19 | 5 G
SDP
Catalog No. 5000446053
Encompass_Preferred
2,4-Dimethylthiazole-5-carboxylic acid is a chemical building block used in various synthetic processes. It appears as a solid with a white to off-white color.
  • Solid appearance
  • White to off-white color
  • Store as powder: -20°C for 3 years, 4°C for 2 years
  • Store in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-46053 Supplier Medchemexpress LLC Supplier No. HYW0022555G
2,4-Dimethylthiazole-5-carboxylic acid is a chemical building block used in various synthetic processes. It appears as a solid with a white to off-white color.
  • Solid appearance
  • White to off-white color
  • Store as powder: -20°C for 3 years, 4°C for 2 years
  • Store in solvent: -80°C for 6 months, -20°C for 1 month