Learn More
Medchemexpress LLC 2,4-Dimethylthiazole-5-carboxylic acid | 53137-27-2 | MFCD00052943 | 157.19 | 500 MG
SDP
Catalog No. 5000447637
Encompass_Preferred
2,4-Dimethylthiazole-5-carboxylic acid is a versatile building block utilized in chemical synthesis, particularly for drug discovery and the development of pharmaceuticals and agrochemicals. Its unique thiazole structure imparts significant functionality, enabling the modification of lead compounds and the enhancement of biological activity. It is suitable for research use only.
  • Used as a heterocyclic building block
  • Enhances biological activity in compounds
  • Important in pharmaceutical and agrochemical synthesis
  • Applicable in the flavoring and fragrance industries
  • Contributes to advancements in medicinal chemistry and agricultural science
Catalog No. 50-004-47637 Supplier Medchemexpress LLC Supplier No. HYW002255500MG
2,4-Dimethylthiazole-5-carboxylic acid is a versatile building block utilized in chemical synthesis, particularly for drug discovery and the development of pharmaceuticals and agrochemicals. Its unique thiazole structure imparts significant functionality, enabling the modification of lead compounds and the enhancement of biological activity. It is suitable for research use only.
  • Used as a heterocyclic building block
  • Enhances biological activity in compounds
  • Important in pharmaceutical and agrochemical synthesis
  • Applicable in the flavoring and fragrance industries
  • Contributes to advancements in medicinal chemistry and agricultural science