Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitro-1-naphthol 98.0+%, TCI America™
SDP

Catalog No. D083925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D083925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D083925G Supplier TCI America Supplier No. D083925G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 605-69-6
Molecular Formula C10H6N2O5
Molecular Weight (g/mol) 234.167
MDL Number MFCD00065352
InChI Key FFRBMBIXVSCUFS-UHFFFAOYSA-N
Synonym 2,4-dinitro-1-naphthol, martius yellow, golden yellow, saffron yellow, manchester yellow, naphthylene yellow, 2,4-dinitronaphthol, martinsgelb, maritus yellow, 1-naphthalenol, 2,4-dinitro
PubChem CID 11802
ChEBI CHEBI:67128
SMILES C1=CC=C2C(=C1)C(=CC(=C2O)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 139°C
Color Yellow
Quantity 25 g
UN Number 3077
Formula Weight 234.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitro-1-naphthol
RTECSNumber QL3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.