Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitroaniline, MilliporeSigma™ Supelco™

Catalog No. 111016734 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6734 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-6734 Supplier MilliporeSigma Supelco Supplier No. 45963100MG
Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 97-02-9
Flash Point 224°C
Physical Form Neat
Linear Formula (O2 N)2C6H3 NH2
MDL Number MFCD00007151
Melting Point 176°C to 178°C
Type Analytical Standard
Quantity 100 mg
Formula Weight 183.12
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.