Learn More
Cayman Chemical 2 4-Dinitrobenzenesulfonic Ac
SDP
Supplier:  Cayman Chemical 4263025 G
Encompass_Preferred
A hapten; induces the proliferation of primary lymphocytes and epithelial cells isolated from 2,4-DNCB-sensitized guinea pigs at 25, 50, or 100 ppm; inhibits 2,4-DNCB-induced contact sensitivity in 2,4-DNCB-sensitized guinea pigs at 600 mg/kg; has been used to induce ulcerative colitis in rats
Catalog No. 50-294-3930