Learn More
Cayman Chemical 2,4-Dinitrobenzenesulfonic Acid (sodium salt), Purity: ≥98%, CAS: 885-62-1, 10 g
SDP
Catalog No. 502945200
Encompass_Preferred
A hapten; induces the proliferation of primary lymphocytes and epithelial cells isolated from 2,4-DNCB-sensitized guinea pigs at 25, 50, or 100 ppm; inhibits 2,4-DNCB-induced contact sensitivity in 2,4-DNCB-sensitized guinea pigs at 600 mg/kg; has been used to induce ulcerative colitis in rats
Catalog No. 50-294-5200 Supplier Cayman Chemical Supplier No. 4263010 G
A hapten; induces the proliferation of primary lymphocytes and epithelial cells isolated from 2,4-DNCB-sensitized guinea pigs at 25, 50, or 100 ppm; inhibits 2,4-DNCB-induced contact sensitivity in 2,4-DNCB-sensitized guinea pigs at 600 mg/kg; has been used to induce ulcerative colitis in rats