Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrofluorobenzene, 98%

Catalog No. AC117030250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117030250 25 g Glass bottle
AC117030050 5 g Glass bottle
AC117031000 100 g Glass bottle
AC117035000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC117030250 Supplier Thermo Scientific Chemicals Supplier No. 117030250
Only null left
Add to Cart

CAS: 70-34-8 | C6H3FN2O4 | 186.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-34-8
Molecular Formula C6H3FN2O4
Molecular Weight (g/mol) 186.1
MDL Number MFCD00007056
InChI Key LOTKRQAVGJMPNV-UHFFFAOYSA-N
Synonym 2,4-dinitrofluorobenzene, dinitrofluorobenzene, fdnb, dnfb, 2,4-dinitro-1-fluorobenzene, sanger's reagent, fluoro-2,4-dinitrobenzene, 2,4-dinitrobenzenefluoride, 2,4-dinitrophenyl fluoride, fluorodinitrobenzene
PubChem CID 6264
ChEBI CHEBI:53049
IUPAC Name 1-fluoro-2,4-dinitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23°C to 26°C
Color Brown to Yellow
Density 1.4800g/mL
Boiling Point 296°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5680 to 1.57
Linear Formula (O2N)2C6H3F
Quantity 25 g
Beilstein 05,262
Fieser 01,321; 02,175; 06,233
Specific Gravity 1.48
Merck Index 15,4204
Solubility Information Solubility in water: 400mg/l (25°C). Other solubilities: soluble in ether,benzene,propylene glycol,and hot alcohol
Formula Weight 186.1
Percent Purity 98%
Physical Form Liquid or Low Melting Crystals
Chemical Name or Material 2, 4-Dinitrofluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause an allergic skin reaction.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic if swallowed.
Suspected of causing cancer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-734-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.