Learn More
Sigma Aldrich Fine Chemicals Biosciences 2,4-Dinitrophenylhydrazine, 119-26-6, MFCD00007578, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D199303100G
Encompass
Linear Formula: (O2N)2C6H3NHNH2, Molecular Weight: 198.14, 97%, MP: 197-200 °C.
Catalog No. NC9898176
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.