Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenylhydrazine, moist solid, contains min. 30% water

Catalog No. AC117065000
Change view
Click to view available options
Quantity:
25 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117065000 500 g Glass bottle
AC117060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC117065000 Supplier Thermo Scientific Chemicals Supplier No. 117065000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

CAS: 119-26-6 | C6H6N4O4 | 198.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-26-6 , 7732-18-5
Molecular Formula C6H6N4O4
MDL Number MFCD00007578
InChI Key HORQAOAYAYGIBM-UHFFFAOYSA-N
Synonym 2,4-dinitrophenyl hydrazine, 2,4-dnph, hydrazine, 2,4-dinitrophenyl, 1-hydrazino-2,4-dinitrobenzene, dnph, brady's reagent, 2,4-dinitrofenylhydrazin, unii-1n39kd7qpj, ccris 3140, 2,4-dnp hydrazine
PubChem CID 3772977
ChEBI CHEBI:66932
IUPAC Name (2,4-dinitrophenyl)hydrazine

Specifications

Melting Point 198°C to 204°C
Color Brown-Orange to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (O2N)2C6H3NHNH2
Quantity 500 g
Beilstein 15, 489
Fieser 01,330; 02,176
Merck Index 15, 3311
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NN
Molecular Weight (g/mol) 198.138
Formula Weight 198.14
Percent Purity ≥98.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 2, 4-Dinitrophenylhydrazine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Flammable solid.
Suspected of causing cancer.
Harmful if swallowed.
Explosive when dry.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-309-3
RTECSNumber MV3325000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.