Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenylhydrazine, Spectrum™ Chemical
SDP

Catalog No. 18600087 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
10 kg
50 kg
Packaging:
Fiber Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-087 50 kg Fiber Drum
18-600-085 25 g Fiber Drum
18-600-089 500 g Fiber Drum
18-600-088 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-600-087 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DI14950KGBL
Only null left
Add to Cart
Add to Cart
Request a Quote

DI149, 119-26-6, C6H6N4O4

Spectrum™ Chemical 2,4-Dinitrophenylhydrazine is a chemical compound that is usually supplied as a wet red-to-orange solid that is shock sensitive. It is used in Brady′s test to detect the carbonyl functionality of an aldehyde or ketone functional group.

Chemical Identifiers

CAS 119-26-6
Molecular Formula C6H6N4O4
Molecular Weight (g/mol) 198.14
InChI Key HORQAOAYAYGIBM-UHFFFAOYSA-N
IUPAC Name (2,4-dinitrophenyl)hydrazine
SMILES NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Max % 100%
Packaging Fiber Drum
Quantity 50 kg
UN Number UN1325
Percent Purity 65 to 70%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.