Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitrophenylsulfenyl Chloride 96.0+%, TCI America™
SDP

Catalog No. D14635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14635G Supplier TCI America Supplier No. D14635G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 528-76-7
Molecular Formula C6H3ClN2O4S
Molecular Weight (g/mol) 234.61
MDL Number MFCD00007129
InChI Key GPXDNWQSQHFKRB-UHFFFAOYSA-N
Synonym 2,4-Dinitrobenzenesulfenyl Chloride
PubChem CID 68251
IUPAC Name (2,4-dinitrophenyl) thiohypochlorite
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])SCl

Specifications

Melting Point 96°C
Color Yellow
Quantity 5 g
UN Number 2925
Formula Weight 234.61
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dinitrophenylsulfenyl Chloride
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.