Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diphenyl-4-methyl-1-pentene 95.0+%, TCI America™
SDP

Catalog No. D2497100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2497100ML 100 mL
D249725ML 25 mL
D2497500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D2497100ML Supplier TCI America Supplier No. D2497100ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6362-80-7
Molecular Formula C18H20
Molecular Weight (g/mol) 236.36
MDL Number MFCD00044027
InChI Key ZOKCNEIWFQCSCM-UHFFFAOYSA-N
Synonym 4-Methyl-2,4-diphenyl-1-pentene
PubChem CID 80715
IUPAC Name (4-methyl-4-phenylpent-1-en-2-yl)benzene
SMILES CC(C)(CC(=C)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 175°C
Quantity 100 mL
UN Number 3082
Formula Weight 236.36
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Diphenyl-4-methyl-1-pentene
EINECSNumber (4)-0852&(4)-0854
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.