Learn More
Medchemexpress LLC 2,4-Disulfamyl-5-trifluoromethylaniline | 654-62-6 | 99.98% | 319.28 | 250 MG
SDP
Catalog No. 5000453284
Encompass_Preferred
DTA, also known as 2,4-Disulfamyl-5-trifluoromethylaniline, is a chemical compound that acts as a cyclic AMP phosphodiesterase inhibitor. It is recognized for its ability to bind to erythrocyte carbonic anhydrase.
  • Acts as a cyclic AMP phosphodiesterase inhibitor.
  • Binds to erythrocyte carbonic anhydrase.
Catalog No. 50-004-53284 Supplier Medchemexpress LLC Supplier No. 250MGHYW011293
DTA, also known as 2,4-Disulfamyl-5-trifluoromethylaniline, is a chemical compound that acts as a cyclic AMP phosphodiesterase inhibitor. It is recognized for its ability to bind to erythrocyte carbonic anhydrase.
  • Acts as a cyclic AMP phosphodiesterase inhibitor.
  • Binds to erythrocyte carbonic anhydrase.