Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-ethoxyphenyl)-3-[4-(methylsulfonyl)phenyl]-pyrazolo[1,5-b]pyridazine | 221148-46-5 | 99.7% | 5 MG
SDP

Catalog No. 5000233169
Encompass_Preferred

GW-406381 is a highly selective cyclooxygenase-2 (COX-2) inhibitor used as a research reagent in preclinical pharmacology and pain models. It attenuates spontaneous ectopic discharge in sural nerves following chronic constriction injury in rats and is provided as a high-purity solid for laboratory use.

  • Selective COX-2 inhibitor.
  • Used in preclinical pain and neurobiology studies.
  • High purity (≈99.7%).
  • Molecular weight 393.46 g/mol.
  • Chemical formula C21H19N3O3S.
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported with ≥2.5 mg/mL solubility.
  • Powder storage: -20°C for long-term stability.
  • Recommended DMSO stock concentrations: 1 mM, 5 mM, and 10 mM (mass-to-volume guidance available).

Catalog No. 50-002-33169 Supplier Medchemexpress LLC Supplier No. HY1193045MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GW-406381 is a highly selective cyclooxygenase-2 (COX-2) inhibitor used as a research reagent in preclinical pharmacology and pain models. It attenuates spontaneous ectopic discharge in sural nerves following chronic constriction injury in rats and is provided as a high-purity solid for laboratory use.

  • Selective COX-2 inhibitor.
  • Used in preclinical pain and neurobiology studies.
  • High purity (≈99.7%).
  • Molecular weight 393.46 g/mol.
  • Chemical formula C21H19N3O3S.
  • Soluble in DMSO (100 mg/mL); in vivo formulations reported with ≥2.5 mg/mL solubility.
  • Powder storage: -20°C for long-term stability.
  • Recommended DMSO stock concentrations: 1 mM, 5 mM, and 10 mM (mass-to-volume guidance available).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.