Learn More
Medchemexpress LLC 2-[(4-fluorobenzyl)amino]ethanol | 22116-33-2 | MFCD04560903 | 95.0% | 169.20 g/mol | C9H12FNO | 500 MG
SDP
Catalog No. 5000280565
Encompass_Preferred
2-[(4-fluorobenzyl)amino]ethanol is an organic building block and drug intermediate used in medicinal chemistry and small-molecule synthesis. The compound is a freestanding amine-alcohol (free base) used to introduce a 4-fluorobenzylaminoethyl fragment into target molecules. Typical analytical values include formula C9H12FNO, molecular weight 169.20 g/mol, and supplier-reported purity around 95.0%.
  • Useful building block for medicinal chemistry synthesis.
  • Introduces a 4-fluorobenzylaminoethyl motif for target elaboration.
  • Free-base amine-alcohol form suitable for common synthetic transformations.
  • Typical purity around 95.0% for synthetic applications.
  • Molecular weight 169.20 g/mol and formula C9H12FNO for formulation calculations.
Catalog No. 50-002-80565 Supplier Medchemexpress LLC Supplier No. HYW157028500MG
2-[(4-fluorobenzyl)amino]ethanol is an organic building block and drug intermediate used in medicinal chemistry and small-molecule synthesis. The compound is a freestanding amine-alcohol (free base) used to introduce a 4-fluorobenzylaminoethyl fragment into target molecules. Typical analytical values include formula C9H12FNO, molecular weight 169.20 g/mol, and supplier-reported purity around 95.0%.
  • Useful building block for medicinal chemistry synthesis.
  • Introduces a 4-fluorobenzylaminoethyl motif for target elaboration.
  • Free-base amine-alcohol form suitable for common synthetic transformations.
  • Typical purity around 95.0% for synthetic applications.
  • Molecular weight 169.20 g/mol and formula C9H12FNO for formulation calculations.