Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Fluorophenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. F07275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07275G 5 g
F07271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07275G Supplier TCI America Supplier No. F07275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58861-53-3
Molecular Formula C11H8FN
Molecular Weight (g/mol) 173.19
MDL Number MFCD06201382
InChI Key MHWIDTQQBWGUCD-UHFFFAOYSA-N
PubChem CID 100868
IUPAC Name 2-(4-fluorophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=CC=C(C=C2)F

Specifications

Melting Point 40°C
Boiling Point 119°C
Quantity 5 g
Formula Weight 173.19
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.