Learn More
Medchemexpress LLC 2,4-Hexadienedioic acid, (2E,4E)- | 3588-17-8 | 99.9% | 142.11 | 1 ML
SDP
Catalog No. 5000437246
Encompass_Preferred
trans-trans-Muconic acid is a urinary metabolite of benzene, frequently utilized as a biomarker for human exposure to benzene. It is also known to be mutagenic in E. coli assays.
  • Analytical standard for research and analytical applications.
  • Used as a biomarker of exposure to benzene in humans.
  • Mutagenic in E. coli assay.
  • Human endogenous metabolite.
  • Microbial metabolite.
Catalog No. 50-004-37246 Supplier Medchemexpress LLC Supplier No. HY11324710MM/1ML
trans-trans-Muconic acid is a urinary metabolite of benzene, frequently utilized as a biomarker for human exposure to benzene. It is also known to be mutagenic in E. coli assays.
  • Analytical standard for research and analytical applications.
  • Used as a biomarker of exposure to benzene in humans.
  • Mutagenic in E. coli assay.
  • Human endogenous metabolite.
  • Microbial metabolite.