Learn More
Medchemexpress LLC 2-(4-hydroxy-3-methoxyphenyl)-benzothiazole | 36341-25-0 | MFCD00030231 | 99.3% | 257.31 | C14H11NO2S | 10 MG
SDP
Catalog No. 5000295247
Encompass_Preferred
YL-109 is a small-molecule antitumor research compound that induces expression of the carboxyl terminus of Hsp70-interacting protein (CHIP) via aryl hydrocarbon receptor signaling. It inhibits breast cancer cell proliferation, motility, and invasiveness in vitro and has shown tumor growth and metastasis suppression in vivo. Intended for research use only.
  • Induces CHIP expression through aryl hydrocarbon receptor signaling.
  • Inhibits breast cancer cell proliferation, motility, and invasiveness in vitro.
  • Demonstrated tumor growth and metastasis suppression in vivo.
  • High purity: 99.3%.
  • Molecular formula C14H11NO2S and molecular weight 257.31 g/mol.
  • Available as a solid and as a 10 mM solution in DMSO for cell-based studies.
Catalog No. 50-002-95247 Supplier Medchemexpress LLC Supplier No. HY1861910MG
YL-109 is a small-molecule antitumor research compound that induces expression of the carboxyl terminus of Hsp70-interacting protein (CHIP) via aryl hydrocarbon receptor signaling. It inhibits breast cancer cell proliferation, motility, and invasiveness in vitro and has shown tumor growth and metastasis suppression in vivo. Intended for research use only.
  • Induces CHIP expression through aryl hydrocarbon receptor signaling.
  • Inhibits breast cancer cell proliferation, motility, and invasiveness in vitro.
  • Demonstrated tumor growth and metastasis suppression in vivo.
  • High purity: 99.3%.
  • Molecular formula C14H11NO2S and molecular weight 257.31 g/mol.
  • Available as a solid and as a 10 mM solution in DMSO for cell-based studies.