Learn More
Medchemexpress LLC 2-(4-hydroxyphenyl)-1-(1,3,4,9-tetrahydro-2H-pyrido[3,4-b]indol-2-yl)-ethanone | 1373764-75-0 | 98.3% | 10 MG
SDP
Catalog No. 5000234533
Encompass_Preferred
YH-306 is a small-molecule focal adhesion kinase (FAK) inhibitor used in research to study tumor growth, migration, and metastasis. It shows antitumor activity in colorectal cancer models and is supplied at high purity for biochemical and cellular assays.
  • Small-molecule FAK inhibitor for pathway and oncology research.
  • Demonstrated suppression of colorectal tumor growth and metastasis in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Available in small research pack sizes for screening and dose-response studies.
  • Chemical formula C19H18N2O2; molecular weight 306.36 g/mol.
Catalog No. 50-002-34533 Supplier Medchemexpress LLC Supplier No. HY12021310MG
YH-306 is a small-molecule focal adhesion kinase (FAK) inhibitor used in research to study tumor growth, migration, and metastasis. It shows antitumor activity in colorectal cancer models and is supplied at high purity for biochemical and cellular assays.
  • Small-molecule FAK inhibitor for pathway and oncology research.
  • Demonstrated suppression of colorectal tumor growth and metastasis in preclinical studies.
  • High purity suitable for biochemical and cellular assays.
  • Available in small research pack sizes for screening and dose-response studies.
  • Chemical formula C19H18N2O2; molecular weight 306.36 g/mol.