Learn More
Medchemexpress LLC 2-(4-hydroxyphenyl)-1-(1,3,4,9-tetrahydro-2H-pyrido[3,4-b]indol-2-yl)-ethanone | 1373764-75-0 | 98.3% | 50 MG
SDP
Catalog No. 5000232155
Encompass_Preferred
YH-306 is a small-molecule focal adhesion kinase (FAK) inhibitor with reported antitumor activity. It suppresses colorectal tumor growth and metastasis by inhibiting FAK signaling, reducing cell migration and invasion, and promoting apoptosis.
  • faK pathway inhibition reduces tumor cell migration and invasion.
  • demonstrated antitumor activity in colorectal cancer models.
  • modulates multiple signaling proteins including c-Src, paxillin, PI3K, and Rac1.
  • downregulates MMP2 and MMP9 expression to limit metastasis.
  • supplied as a research reagent, soluble in DMSO for experimental use.
Catalog No. 50-002-32155 Supplier Medchemexpress LLC Supplier No. HY12021350MG
YH-306 is a small-molecule focal adhesion kinase (FAK) inhibitor with reported antitumor activity. It suppresses colorectal tumor growth and metastasis by inhibiting FAK signaling, reducing cell migration and invasion, and promoting apoptosis.
  • faK pathway inhibition reduces tumor cell migration and invasion.
  • demonstrated antitumor activity in colorectal cancer models.
  • modulates multiple signaling proteins including c-Src, paxillin, PI3K, and Rac1.
  • downregulates MMP2 and MMP9 expression to limit metastasis.
  • supplied as a research reagent, soluble in DMSO for experimental use.