Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(4-Hydroxyphenyl)ethanol

Catalog No. 501855067
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-185-5067 25 g
50-185-5068 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-185-5067 Supplier Sigma Aldrich Supplier No. 18825525G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(4-Hydroxyphenyl)ethanol
TRUSTED_SUSTAINABILITY

Specifications

CAS 501-94-0
Melting Point 89°C to 92°C (lit.)
Molecular Formula C8H10O2
Linear Formula HOC6H4CH2CH2OH
MDL Number MFCD00002902
Quantity 25 g
Synonym 4-Hydroxyphenethyl alcohol; Tyrosol; p-HPEA
Molecular Weight (g/mol) 138.16
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.