Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Hydroxyphenyl)ethanol, >99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014348 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4348 100 mg
11-101-4349 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4348 Supplier MilliporeSigma Supelco Supplier No. 79058100MGF
Add to Cart
Add to Cart

2-(4-Hydroxyphenyl)ethanol is a phenolic compound present in olives, which can exhibit a wide range of health benefits and pharmacological properties like anti-inflammatory, antiviral, anti-tumor and cardioprotection activity.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 501-94-0
Physical Form Neat
Linear Formula HOC6H4 CH2 CH2 OH
MDL Number MFCD00002902
Melting Point 89°C to 92°C; 90°C to 93°C
Synonym 4-Hydroxyphenethyl alcohol; Tyrosol; p-HPEA
Type Analytical Standard
Quantity 100 mg
Formula Weight 138.16
Percent Purity >99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.