Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-hydroxyphenyl)propionic acid | 938-96-5 | MFCD00128847 | 98.7% | 166.17 g/mol | C9H10O3 | 500 MG
SDP

Catalog No. 5000210388
Encompass_Preferred

2-(4-hydroxyphenyl)propionic acid is an aromatic carboxylic acid supplied as a research reagent. It is reported to be convertible into R and S glucosyl esters and glucosides by root culture of P. ginseng (Pg-1). The material is a solid with reported purity of 98.66%.

  • Purity 98.7% by supplier
  • Molecular weight 166.17 g/mol
  • Chemical formula C9H10O3
  • CAS number 938-96-5
  • Offered in a 500 MG pack (other sizes available)
  • Suitable for research applications and synthetic transformations

Catalog No. 50-002-10388 Supplier Medchemexpress LLC Supplier No. HYW067425500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-hydroxyphenyl)propionic acid is an aromatic carboxylic acid supplied as a research reagent. It is reported to be convertible into R and S glucosyl esters and glucosides by root culture of P. ginseng (Pg-1). The material is a solid with reported purity of 98.66%.

  • Purity 98.7% by supplier
  • Molecular weight 166.17 g/mol
  • Chemical formula C9H10O3
  • CAS number 938-96-5
  • Offered in a 500 MG pack (other sizes available)
  • Suitable for research applications and synthetic transformations

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.