Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(4-hydroxyphenyl)propionic acid | 938-96-5 | MFCD00128881 | 98.7% | 166.17 g/mol | C9H10O3
SDP

Catalog No. 5000208892
Encompass_Preferred

2-(4-hydroxyphenyl)propionic acid is an aromatic carboxylic acid used as a research reagent and synthetic intermediate in organic and natural products chemistry. It has been reported to be converted into R and S glucosyl esters and glucosides by Panax ginseng root culture. Typical analytical data reported by suppliers include CAS 938-96-5, formula C9H10O3, molecular weight 166.17 g/mol, and reported purity about 98.7%.

  • Aromatic carboxylic acid used as a research reagent.
  • Reported conversion to R and S glucosyl esters and glucosides by P. ginseng root culture.
  • CAS number 938-96-5 for unambiguous identification.
  • Chemical formula C9H10O3 and molecular weight 166.17 g/mol.
  • High reported purity suitable for research applications (~98.7%).

Catalog No. 50-002-08892 Supplier Medchemexpress LLC Supplier No. 250MGHYW067425
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(4-hydroxyphenyl)propionic acid is an aromatic carboxylic acid used as a research reagent and synthetic intermediate in organic and natural products chemistry. It has been reported to be converted into R and S glucosyl esters and glucosides by Panax ginseng root culture. Typical analytical data reported by suppliers include CAS 938-96-5, formula C9H10O3, molecular weight 166.17 g/mol, and reported purity about 98.7%.

  • Aromatic carboxylic acid used as a research reagent.
  • Reported conversion to R and S glucosyl esters and glucosides by P. ginseng root culture.
  • CAS number 938-96-5 for unambiguous identification.
  • Chemical formula C9H10O3 and molecular weight 166.17 g/mol.
  • High reported purity suitable for research applications (~98.7%).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.